Learn More
trans-Diamminedichloroplatinum(II), Pt 64.5% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01047201
Description
trans-Diamminedichloroplatinum(II) acts as a reversible RNA-protein cross-linking agent. It is also used in plating baths. It reacts with DNA after hydrolysis to get both intra and interstrand crosslinks. These crosslinks appear to impair replication and transcription of DNA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
trans-Diamminedichloroplatinum(II) | |
14913-33-8 | |
Crystalline | |
Cl2H6N2Pt | |
MFCD00011623 | |
Soluble in dimethylsulfoxide and dimethylformamide. Slightly soluble in water. | |
N.N.Cl[Pt]Cl | |
300.05 | |
CHEBI:27899 | |
Pt 64.5% min. |
340°C (dec.) | |
Yellow | |
0.25 g | |
PtCl2(NH3)2 | |
trans-dichlorodiamineplatinum ii, azane; dichloroplatinum, h6cl2n2pt, ia call, ddp-h, platinol tn, cl.2h3n.pt, platinum ii chloride diamine, spectrum1502107, cisplatin jp17/usp/inn | |
LXZZYRPGZAFOLE-UHFFFAOYSA-L | |
azane;dichloroplatinum | |
5702198 | |
300.06 | |
Odorless |
Chemical Identifiers
14913-33-8 | |
300.05 | |
LXZZYRPGZAFOLE-UHFFFAOYSA-L | |
5702198 | |
azane;dichloroplatinum |
Cl2H6N2Pt | |
MFCD00011623 | |
trans-dichlorodiamineplatinum ii, azane; dichloroplatinum, h6cl2n2pt, ia call, ddp-h, platinol tn, cl.2h3n.pt, platinum ii chloride diamine, spectrum1502107, cisplatin jp17/usp/inn | |
CHEBI:27899 | |
N.N.Cl[Pt]Cl |
Safety and Handling
GHS H Statement
H334-H341-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H317-H318-H334-H340-H351
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 238-980-9
RTECSNumber : TP2455000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only