missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-Diamminedichloropalladium(II), Premion™, 99.95% (metals basis), Pd 49.9% min

Manufacturer:  Alfa Aesar™ 1103702

 View more versions of this product

Catalog No. AA1103702



Specifications

Specifications

trans-Diamminedichloropalladium(II)
0.5g
(metals basis)
MFCD00011621
diammine dichloropalladium, trans-diamminedichloropalladium ii, transpalladin, cl2pd.2nh3, trans-bisamminedichloropalladium, palladium chloride diamine, trans-dichlorodiammine palladium ii, trans-dichlorodiammine palladium ii, trans-diamminedichloropalladium ii , premion, trans-diamminedichloropalladium ii
Slightly soluble in water; Soluble in NH4OH; Insoluble in chloroform, acetone
N.N.Cl[Pd]Cl
211.382
211.37
99.95%
Pd 49.9% min.
Yellow
14323-43-4
Cl2H6N2Pd
UN3077
Pd 49.9% min.
WIEZTXFTOIBIOC-UHFFFAOYSA-L
azane;dichloropalladium
61732
Crystalline Powder
Premion™
2.5g/mL

Chemical Identifiers

14323-43-4
211.382
WIEZTXFTOIBIOC-UHFFFAOYSA-L
61732
N.N.Cl[Pd]Cl
Cl2H6N2Pd
MFCD00011621
diammine dichloropalladium, trans-diamminedichloropalladium ii, transpalladin, cl2pd.2nh3, trans-bisamminedichloropalladium, palladium chloride diamine, trans-dichlorodiammine palladium ii, trans-dichlorodiammine palladium ii, trans-diamminedichloropalladium ii , premion, trans-diamminedichloropalladium ii
azane;dichloropalladium
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : RT3560000

TSCA : TSCA

Documents