Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-Diamminedichloropalladium(II), Premion™, 99.95% (metals basis), Pd 49.9% min

Chemical Identifiers

Chemical Name or Material trans-Diamminedichloropalladium(II)
CAS 14323-43-4
Purity Grade Premion™
Formula Weight 211.37
MDL Number MFCD00011621
UN Number UN3077
Synonym Dichlorodiamminepalladium(II)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1103704

  SDS
alfa aesar™
1103704
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1103702

  SDS
alfa aesar™
1103702
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1103709

  SDS
alfa aesar™
1103709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material trans-Diamminedichloropalladium(II)
CAS 14323-43-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number RT3560000
TSCA TSCA
Purity Grade Premion™
Physical Form Crystalline Powder
Assay 99.95%
Assay Percent Notes (metals basis)
Formula Weight 211.37
MDL Number MFCD00011621
Density 2.5g/mL
UN Number UN3077
Synonym Dichlorodiamminepalladium(II)
Concentration Pd 49.9% min.
Solubility Information Slightly soluble in water; Soluble in NH4OH; Insoluble in chloroform, acetone
Color Yellow
Quantity 2g
Safety and Handling

Danger

GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.