Please login to your online account to display your discounted pricing

trans-Crotyl bromide, remainder 3-bromo-1-butene, 85%, ACROS Organics™


Quantity 25mL

Manufacturer: thermo fisher scientific  292490250

 View more versions of this product

Catalog No. AC292490250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥80%
Physical Form Liquid
Formula Weight 135
Quantity 25mL
Color Clear light yellow to yellow-brown
Beilstein 01, 205
TSCA TSCA
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material trans-Crotyl bromide, remainder 3-bromo-1-butene
Name Note 85%
Assay Percent Range 85%
MDL Number MFCD00000248
CAS 29576-14-5
Infrared Spectrum Authentic
Refractive Index 1.4785 to 1.4805 (20°C, 589nm)
Molecular Formula C4H7Br
Linear Formula CH3CH=CHCH2Br
Synonym trans-1-Bromo-2-butene
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material trans-Crotyl bromide, remainder 3-bromo-1-butene
Name Note 85%
CAS 29576-14-5
Assay ≥80%
Assay Percent Range 85%
Molecular Formula C4H7Br
Linear Formula CH3CH=CHCH2Br
Formula Weight 135
MDL Number MFCD00000248
Synonym trans-1-Bromo-2-butene
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents