Please login to your online account to display your discounted pricing

trans-Crotyl bromide, remainder 3-bromo-1-butene, 85%, ACROS Organics™

$149.76 - $149.76

Chemical Identifiers

Chemical Name or Material trans-Crotyl bromide, remainder 3-bromo-1-butene
Name Note 85%
CAS 29576-14-5
Assay 80% min.
Assay Percent Range 85%
Molecular Formula C4H7Br
Linear Formula CH3CH=CHCH2Br
Formula Weight 135.00g/mol
MDL Number MFCD00000248
Synonym trans-1-Bromo-2-butene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC292490250

  SDS
thermo fisher scientific
292490250
25mL Each for $149.76
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4785 to 1.4805 (20°C, 589nm)
Color Clear light yellow to yellow-brown
Quantity 25mL
Chemical Name or Material trans-Crotyl bromide, remainder 3-bromo-1-butene
Name Note 85%
CAS 29576-14-5
Physical Form Clear light yellow to yellow-brown liquid
Assay 80% min.
Assay Percent Range 85%
Molecular Formula C4H7Br
Linear Formula CH3CH=CHCH2Br
Formula Weight 135.00g/mol
MDL Number MFCD00000248
Beilstein 01, 205
Synonym trans-1-Bromo-2-butene
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.