missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-Cinnamoyl chloride, 97%

Manufacturer:  Alfa Aesar™ A1201622

 View more versions of this product

Catalog No. AAA1201622



Description

Description

Applications
The derivative of this compound, trans-Cinnamoylimidazole is involved in the synthesis of trans-Cinnamoyl-Papain.

Solubility
Reacts with water.

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases. Moisture sensitive.
Specifications

Specifications

trans-Cinnamoyl chloride
Pungent
35°C to 37°C
17082-09-6
MFCD00000732
606265
cinnamoyl chloride, cinnamic acid chloride, cinnamic chloride, trans-3-phenylacryloyl chloride, 2-propenoyl chloride, 3-phenyl, cinnamoylchloride, e-cinnamoyl chloride, trans-cinnamoyl chloride, 2e-3-phenylprop-2-enoyl chloride, 3-phenyl-2-propenoyl chloride
C1=CC=C(C=C1)C=CC(=O)Cl
166.604
166.61
>110°C (230°F)
256°C to 258°C
100g
C9H7ClO
UN3261
14,2301
WOGITNXCNOTRLK-VOTSOKGWSA-N
(E)-3-phenylprop-2-enoyl chloride
5354261
97%

Chemical Identifiers

17082-09-6
166.604
WOGITNXCNOTRLK-VOTSOKGWSA-N
5354261
C1=CC=C(C=C1)C=CC(=O)Cl
C9H7ClO
MFCD00000732
cinnamoyl chloride, cinnamic acid chloride, cinnamic chloride, trans-3-phenylacryloyl chloride, 2-propenoyl chloride, 3-phenyl, cinnamoylchloride, e-cinnamoyl chloride, trans-cinnamoyl chloride, 2e-3-phenylprop-2-enoyl chloride, 3-phenyl-2-propenoyl chloride
(E)-3-phenylprop-2-enoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

SDS Documents