missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-Cinnamaldehyde, ≥98%, Alfa Aesar™

Manufacturer:  Alfa Aesar A146890E

 View more versions of this product

Catalog No. AAA146890E



Description

Description

Applications
trans-Cinnamaldehyde is used in the flavor and perfume industry. It is also used in medicine. It reacts with glutathione to get an adduct 1′-(glutathion-S-yl)-dihydrocinnamaldehyde. It is used to prepare cinnamylidene-bisacetamide by reacting with acetamide. Further, it inhibits xanthine oxidase.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases. Air sensitive.
Specifications

Specifications

trans-Cinnamaldehyde
Characteristic
-8°C
14371-10-9
C9H8O
1071571
cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal
C1=CC=C(C=C1)C=CC=O
132.162
CHEBI:16731
Liquid
131°C (268°F)
250°C to 252°C
2500g
≥98%
MFCD00007000
14,2297
KJPRLNWUNMBNBZ-QPJJXVBHSA-N
(E)-3-phenylprop-2-enal
637511
132.16
≥98%

Chemical Identifiers

14371-10-9
132.162
KJPRLNWUNMBNBZ-QPJJXVBHSA-N
637511
(E)-3-phenylprop-2-enal
C9H8O
MFCD00007000
cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal
CHEBI:16731
C1=CC=C(C=C1)C=CC=O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P280g-P262-P305+P351+P338
Wear protective gloves.
Do not get in eyes, on skin, or on clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : GD6475000

storageNote1 : Air sensitive

Documents