missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-Chalcone, 97%

Manufacturer:  Alfa Aesar™ A1473436

 View more versions of this product

Catalog No. AAA1473436



Description

Description

Applications
Trans chalcone is used to prevent lung and forestomach cancer. It induces programmed cell death due to reduced mitochondrial transmembrane potential in Arabidopsis thaliana roots.

Solubility
Soluble in chloroform, ether, benzene, and ethanol (slightly). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

trans-Chalcone
55°C to 57°C
614-47-1
MFCD00003082
14,2037
DQFBYFPFKXHELB-VAWYXSNFSA-N
(E)-1,3-diphenylprop-2-en-1-one
637760
208.26
208°C (25mmHg)
500g
C15H12O
509985
chalcone, trans-chalcone, e-chalcone, benzalacetophenone, chalkone, benzylideneacetophenone, phenyl styryl ketone, cinnamophenone, 2-benzalacetophenone, 2-benzylideneacetophenone
C1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2
208.26
CHEBI:48965
97%

Chemical Identifiers

614-47-1
208.26
DQFBYFPFKXHELB-VAWYXSNFSA-N
637760
(E)-1,3-diphenylprop-2-en-1-one
C15H12O
MFCD00003082
chalcone, trans-chalcone, e-chalcone, benzalacetophenone, chalkone, benzylideneacetophenone, phenyl styryl ketone, cinnamophenone, 2-benzalacetophenone, 2-benzylideneacetophenone
CHEBI:48965
C1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UD5576750

Documents