missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-Bis(benzonitrile)dichloropalladium(II), Pd 27.1%

Manufacturer:  Alfa Aesar™ 1000603

 View more versions of this product

Catalog No. AA1000603



Specifications

Specifications

trans-Bis(benzonitrile)dichloropalladium(II)
1g
C14H10Cl2N2Pd
MFCD00013123
bis benzonitrile palladium chloride, bis benzonitrile palladium ii chloride, trans-bis benzonitrile palladium ii chloride, bis benzonitrile palladium ii chloride, 500mg, palladium 2+ bis benzonitrile dichloride
Soluble in acetone, chloroform
C1=CC=C(C=C1)C#N.C1=CC=C(C=C1)C#N.[Cl-].[Cl-].[Pd+2]
383.568
383.57
White
14220-64-5
PdCl2[(C6H5)(CN)]2
UN3439
Pd 27.1%
WXNOJTUTEXAZLD-UHFFFAOYSA-L
benzonitrile;palladium(2+);dichloride
10271322
Powder

Chemical Identifiers

14220-64-5
383.568
WXNOJTUTEXAZLD-UHFFFAOYSA-L
10271322
C1=CC=C(C=C1)C#N.C1=CC=C(C=C1)C#N.[Cl-].[Cl-].[Pd+2]
C14H10Cl2N2Pd
MFCD00013123
bis benzonitrile palladium chloride, bis benzonitrile palladium ii chloride, trans-bis benzonitrile palladium ii chloride, bis benzonitrile palladium ii chloride, 500mg, palladium 2+ bis benzonitrile dichloride
benzonitrile;palladium(2+);dichloride
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents