Please login to your online account to display your discounted pricing

(±)-trans-ACPD, Tocris


Group I/group II mGlu agonist

Quantity 10mg

Manufacturer: r & d systems  RDS018710

 View more versions of this product

Catalog No. 018710

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 173.17
Molecular Formula C7H11NO4
CAS 67684-64-4
Chemical Name or Material (±)-trans-ACPD
Assay Percent Range >99%
Synonym (±)-1-Aminocyclopentane-trans-1, 3-dicarboxylic acid
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material (±)-trans-ACPD
Assay Percent Range >99%
Molecular Formula C7H11NO4
Formula Weight 173.17
Synonym (±)-1-Aminocyclopentane-trans-1, 3-dicarboxylic acid
Accessories

Store at Room Temperature