missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-9-Octadecenol 99.0+%, TCI America™

Manufacturer:  TCI America O02285G

 View more versions of this product

Catalog No. O02285G



Specifications

Specifications

trans-9-Octadecenol
White
5g
C18H36O
Elaidyl Alcohol
CCCCCCCCC=CCCCCCCCCO
268.485
268.49
≥99.0% (GC)
333°C
37°C
506-42-3
MFCD00063218
ALSTYHKOOCGGFT-MDZDMXLPSA-N
(E)-octadec-9-en-1-ol
5367665
Crystal-Powder at 20°C

Chemical Identifiers

506-42-3
268.485
ALSTYHKOOCGGFT-MDZDMXLPSA-N
5367665
CCCCCCCCC=CCCCCCCCCO
C18H36O
MFCD00063218
Elaidyl Alcohol
(E)-octadec-9-en-1-ol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0258

TSCA : No

Recommended Storage : Refrigerator

Documents