missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-4-(Trifluoromethyl)cyclohexanecarboxylic Acid 98.0+%, TCI America™

$87.51 - $274.29

Chemical Identifiers

CAS 133261-33-3
Molecular Formula C8H11F3O2
Molecular Weight (g/mol) 196.169
InChI Key LMEAZIIFLVDISW-UHFFFAOYSA-N
Synonym 4-trifluoromethyl cyclohexanecarboxylic acid, trans-4-trifluoromethyl cyclohexanecarboxylic acid, cis-4-trifluoromethyl cyclohexanecarboxylic acid, 4-trifluoromethyl cyclohexane-1-carboxylic acid, 1r,4r-4-trifluoromethyl cyclohexanecarboxylic acid, trans-4-trifluoromethyl cyclohexane-carboxylicacid, trans-4-trifluoromethyl cyclohexane-carboxylic acid, trans-4-trifluoromethylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-trifluoromethyl, 4-trifluoromethyl-cyclohexanecarboxylic acid
PubChem CID 2779182
IUPAC Name 4-(trifluoromethyl)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T24241G
View Documents Promotion Details
TCI America
T24241G
1 g
Each for $87.51
 
T24245G
View Documents Promotion Details
TCI America
T24245G
5 g
Each for $274.29
 
Description

Description

Specifications

Chemical Identifiers

133261-33-3
196.169
4-trifluoromethyl cyclohexanecarboxylic acid, trans-4-trifluoromethyl cyclohexanecarboxylic acid, cis-4-trifluoromethyl cyclohexanecarboxylic acid, 4-trifluoromethyl cyclohexane-1-carboxylic acid, 1r,4r-4-trifluoromethyl cyclohexanecarboxylic acid, trans-4-trifluoromethyl cyclohexane-carboxylicacid, trans-4-trifluoromethyl cyclohexane-carboxylic acid, trans-4-trifluoromethylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-trifluoromethyl, 4-trifluoromethyl-cyclohexanecarboxylic acid
4-(trifluoromethyl)cyclohexane-1-carboxylic acid
C8H11F3O2
LMEAZIIFLVDISW-UHFFFAOYSA-N
2779182
C1CC(CCC1C(=O)O)C(F)(F)F

Specifications

133261-33-3
White-Yellow
1 g
LMEAZIIFLVDISW-UHFFFAOYSA-N
4-(trifluoromethyl)cyclohexane-1-carboxylic acid
2779182
≥98.0% (GC,T)
trans-4-(Trifluoromethyl)cyclohexanecarboxylic Acid
154°C
C8H11F3O2
4-trifluoromethyl cyclohexanecarboxylic acid, trans-4-trifluoromethyl cyclohexanecarboxylic acid, cis-4-trifluoromethyl cyclohexanecarboxylic acid, 4-trifluoromethyl cyclohexane-1-carboxylic acid, 1r,4r-4-trifluoromethyl cyclohexanecarboxylic acid, trans-4-trifluoromethyl cyclohexane-carboxylicacid, trans-4-trifluoromethyl cyclohexane-carboxylic acid, trans-4-trifluoromethylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-trifluoromethyl, 4-trifluoromethyl-cyclohexanecarboxylic acid
C1CC(CCC1C(=O)O)C(F)(F)F
196.169
196.17
Crystalline Powder
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'