missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-tert-Pentylcyclohexanol, 97%

$254.31 - $254.31

Chemical Identifiers

CAS 20698-30-0
Molecular Formula C11H22O
Formula Weight 170.3
MDL Number MFCD00067157
Synonym trans-4-tert-Amylcyclohexanol
Chemical Name or Material trans-4-tert-Pentylcyclohexanol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2147906 SDS Alfa Aesar™
B2147906
5g Each for $254.31
AAB2147903 SDS Alfa Aesar™
B2147903
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

20698-30-0
170.3
trans-4-tert-Amylcyclohexanol
C11H22O
MFCD00067157
trans-4-tert-Pentylcyclohexanol

Specifications

20698-30-0
24° to 27°C
5g
MFCD00067157
trans-4-tert-Pentylcyclohexanol
154° to 156°C (40mmHg)
C11H22O
170.3
trans-4-tert-Amylcyclohexanol
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning