missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-4-(tert-Butoxycarbonylamino)cyclohexanol, TCI America™

$39.72 - $129.86

Chemical Identifiers

CAS 111300-06-2
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.293
InChI Key DQARDWKWPIRJEH-UHFFFAOYSA-N
Synonym trans-4-boc-aminocyclohexanol, tert-butyl 4-hydroxycyclohexyl carbamate, boc-trans-4-aminocyclohexanol, 4-boc-amino cyclohexanol, tert-butyl trans-4-hydroxycyclohexyl carbamate, tert-butyl n-4-hydroxycyclohexyl carbamate, tert-butyl cis-4-hydroxycyclohexylcarbamate, trans-n-boc-4-amino-cyclohexanol, 4-n-boc-amino-cyclohexanol, tert-butyl cis-4-hydroxycyclohexyl carbamate
PubChem CID 1514287
IUPAC Name tert-butyl N-(4-hydroxycyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B35641G View Documents TCI America
B35641G
1g Each for $39.72
 
B35645G View Documents TCI America
B35645G
5g Each for $129.86
 
Specifications

Chemical Identifiers

111300-06-2
215.293
trans-4-boc-aminocyclohexanol, tert-butyl 4-hydroxycyclohexyl carbamate, boc-trans-4-aminocyclohexanol, 4-boc-amino cyclohexanol, tert-butyl trans-4-hydroxycyclohexyl carbamate, tert-butyl n-4-hydroxycyclohexyl carbamate, tert-butyl cis-4-hydroxycyclohexylcarbamate, trans-n-boc-4-amino-cyclohexanol, 4-n-boc-amino-cyclohexanol, tert-butyl cis-4-hydroxycyclohexyl carbamate
tert-butyl N-(4-hydroxycyclohexyl)carbamate
C11H21NO3
DQARDWKWPIRJEH-UHFFFAOYSA-N
1514287
CC(C)(C)OC(=O)NC1CCC(CC1)O

Specifications

White-Yellow
1g
C11H21NO3
DQARDWKWPIRJEH-UHFFFAOYSA-N
tert-butyl N-(4-hydroxycyclohexyl)carbamate
1514287
Crystal-Powder at 20°C
trans-4-(tert-Butoxycarbonylamino)cyclohexanol
171°C
111300-06-2
trans-4-boc-aminocyclohexanol, tert-butyl 4-hydroxycyclohexyl carbamate, boc-trans-4-aminocyclohexanol, 4-boc-amino cyclohexanol, tert-butyl trans-4-hydroxycyclohexyl carbamate, tert-butyl n-4-hydroxycyclohexyl carbamate, tert-butyl cis-4-hydroxycyclohexylcarbamate, trans-n-boc-4-amino-cyclohexanol, 4-n-boc-amino-cyclohexanol, tert-butyl cis-4-hydroxycyclohexyl carbamate
CC(C)(C)OC(=O)NC1CCC(CC1)O
215.293
215.29
≥98.0% (GC,N)
Safety and Handling

Safety and Handling

TSCA : Yes