Learn More
trans-4-Phenyl-3-buten-2-one, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 164525000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
trans-4-Phenyl-3-buten-2-one | |
1896-62-4 | |
122°C | |
98.5% min. (GC) | |
C10H10O | |
MFCD00008779 | |
07, 364 | |
benzalacetone, benzylideneacetone, methyl styryl ketone, 4-phenylbut-3-en-2-one, acetocinnamone, 4-phenyl-3-buten-2-one, e-4-phenylbut-3-en-2-one, benzylidene acetone, trans-benzalacetone, benzalaceton | |
BWHOZHOGCMHOBV-BQYQJAHWSA-N | |
(E)-4-phenylbut-3-en-2-one | |
637759 | |
146.19 |
99% | |
260°C to 262°C | |
Authentic | |
Glass bottle | |
C6H5CH=CHCOCH3 | |
500g | |
15, 1140 | |
Solubility in water: negligible. Other solubilities: freely soluble in alcohol,benzene,chloroform and,ether | |
CC(=O)C=CC1=CC=CC=C1 | |
146.19 | |
CHEBI:78399 | |
99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice
GHS Signal Word: Warning
EINECSNumber : 217-587-6
RTECSNumber : EN0330050
RUO â Research Use Only