missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-4-Nitrostilbene, TCI America™

$91.67 - $265.84

Chemical Identifiers

CAS 1694-20-8
Molecular Formula C14H11NO2
Molecular Weight (g/mol) 225.247
MDL Number MFCD00053614
InChI Key ZISCOWXWCHUSMH-VOTSOKGWSA-N
PubChem CID 639659
ChEBI CHEBI:377106
IUPAC Name 1-nitro-4-[(E)-2-phenylethenyl]benzene
SMILES C1=CC=C(C=C1)C=CC2=CC=C(C=C2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N05495G View Documents TCI America
N05495G
5g Each for $91.67
 
N054925G View Documents TCI America
N054925G
25g Each for $265.84
 
Specifications

Chemical Identifiers

1694-20-8
225.247
ZISCOWXWCHUSMH-VOTSOKGWSA-N
CHEBI:377106
C1=CC=C(C=C1)C=CC2=CC=C(C=C2)[N+](=O)[O-]
C14H11NO2
MFCD00053614
639659
1-nitro-4-[(E)-2-phenylethenyl]benzene

Specifications

Undesignated
5g
C14H11NO2
ZISCOWXWCHUSMH-VOTSOKGWSA-N
1-nitro-4-[(E)-2-phenylethenyl]benzene
639659
225.25
≥98.0% (GC)
157°C
1694-20-8
MFCD00053614
C1=CC=C(C=C1)C=CC2=CC=C(C=C2)[N+](=O)[O-]
225.247
CHEBI:377106
Crystal-Powder at 20°C
trans-4-Nitrostilbene
Safety and Handling

Safety and Handling

RTECSNumber : WJ6950050

TSCA : No