Learn More
trans-4-Nitrocinnamic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1299022
Description
trans-4-Nitrocinnamic acid, is used as a raw material for saccharine. It is also used as a useful chemical for biological applications. Some other applications include it is used as an organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans-4-Nitrocinnamic acid, is used as a raw material for saccharine. It is also used as a useful chemical for biological applications. Some other applications include it is used as an organic intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases.
Specifications
trans-4-Nitrocinnamic acid | |
∼290°C (decomposition) | |
C9H7NO4 | |
100 g | |
4-nitrocinnamic acid, 3-4-nitrophenyl acrylic acid, p-nitrocinnamic acid, cinnamic acid, p-nitro, 2-propenoic acid, 3-4-nitrophenyl, e-3-4-nitrophenyl acrylic acid, 3-4-nitrophenyl propenoic acid, trans-4-nitrocinnamic acid, ccris 1665, 2e-3-4-nitrophenyl prop-2-enoic acid | |
XMMRNCHTDONGRJ-ZZXKWVIFSA-N | |
193.158 | |
193.16 |
882-06-4 | |
Odorless | |
MFCD00007381 | |
1369659 | |
Insoluble in water. | |
C1=CC(=CC=C1C=CC(=O)O)[N+](=O)[O-] | |
737157 | |
≥98% |
Chemical Identifiers
882-06-4 | |
193.158 | |
XMMRNCHTDONGRJ-ZZXKWVIFSA-N | |
737157 |
C9H7NO4 | |
MFCD00007381 | |
4-nitrocinnamic acid, 3-4-nitrophenyl acrylic acid, p-nitrocinnamic acid, cinnamic acid, p-nitro, 2-propenoic acid, 3-4-nitrophenyl, e-3-4-nitrophenyl acrylic acid, 3-4-nitrophenyl propenoic acid, trans-4-nitrocinnamic acid, ccris 1665, 2e-3-4-nitrophenyl prop-2-enoic acid | |
C1=CC(=CC=C1C=CC(=O)O)[N+](=O)[O-] |
Safety and Handling
GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H341
EINECSNumber : 212-924-3
RTECSNumber : GE0353000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only