missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-n-Propylcyclohexyl trans,trans-4'-n-propylbicyclohexyl-4-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H5513806

 View more versions of this product

Catalog No. AAH5513806



Specifications

Specifications

trans-4-n-Propylcyclohexyl trans,trans-4'-n-propylbicyclohexyl-4-carboxylate
5g
C25H44O2
1,1'-bicyclohexyl-4-carboxylic acid, 4'-propyl-, 4-propylcyclohexyl ester, trans trans trans
CCCC1CCC(CC1)C2CCC(CC2)C(=O)OC3CCC(CC3)CCC
376.625
376.62
168°C to 170°C
83242-83-5
MFCD20527218
ITVBZSWXRMMPFS-UHFFFAOYSA-N
(4-propylcyclohexyl) 4-(4-propylcyclohexyl)cyclohexane-1-carboxylate
550127
97%

Chemical Identifiers

83242-83-5
376.625
ITVBZSWXRMMPFS-UHFFFAOYSA-N
550127
CCCC1CCC(CC1)C2CCC(CC2)C(=O)OC3CCC(CC3)CCC
C25H44O2
MFCD20527218
1,1'-bicyclohexyl-4-carboxylic acid, 4'-propyl-, 4-propylcyclohexyl ester, trans trans trans
(4-propylcyclohexyl) 4-(4-propylcyclohexyl)cyclohexane-1-carboxylate
Safety and Handling

Safety and Handling

EINECSNumber : 280-293-1

TSCA : No

Recommended Storage : Ambient temperatures

Documents