missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl, 97%

Manufacturer:  Alfa Aesar™ H5574803

 View more versions of this product

Catalog No. AAH5574803



Description

Description

Applications
trans-4-n-Propyl-4′-(3,4,5-trifluorophenyl)bicyclohexyl is used to make PDLC mixtures and Switchable Film . It is also mainly used as a liquid crystal monomer, liquid crystal material.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl
1g
C21H29F3
1,2,3-trifluoro-5-trans,trans-4-propyl 1,1-bicyclohexyl-4-yl benzene
FEWMLRARKGRCCE-UHFFFAOYSA-N
1,2,3-trifluoro-5-[4-(4-propylcyclohexyl)cyclohexyl]benzene
11772113
97%
63°C to 64°C
131819-23-3
MFCD09838998
It is soluble in water.
CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C(=C3)F)F)F
338.458
338.46

Chemical Identifiers

131819-23-3
338.458
FEWMLRARKGRCCE-UHFFFAOYSA-N
11772113
CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C(=C3)F)F)F
C21H29F3
MFCD09838998
1,2,3-trifluoro-5-trans,trans-4-propyl 1,1-bicyclohexyl-4-yl benzene
1,2,3-trifluoro-5-[4-(4-propylcyclohexyl)cyclohexyl]benzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents