Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-n-Pentylcyclohexanecarboxylic acid, 98%

$44.76 - $107.43

Chemical Identifiers

Chemical Name or Material trans-4-n-Pentylcyclohexanecarboxylic acid
CAS 38289-29-1
Molecular Formula C12H22O2
Formula Weight 198.3
MDL Number MFCD00009964
Synonym trans-4-n-Pentylcyclohexanecarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2171603

  SDS
alfa aesar™
B2171603
1g Each for $44.76

AAB2171606

  SDS
alfa aesar™
B2171606
5g Each for $107.43
Description & Specifications

Specifications

Formula Weight 198.3
Chemical Name or Material trans-4-n-Pentylcyclohexanecarboxylic acid
MDL Number MFCD00009964
Melting Point 52° to 55°C
Flash Point >110°C (230°F)
Molecular Formula C12H22O2
Synonym trans-4-n-Pentylcyclohexanecarboxylic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 38289-29-1
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.