missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans-4-Methylcyclohexanecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America M178325G
Specifications
trans-4-Methylcyclohexanecarboxylic Acid | |
112°C | |
245°C | |
MFCD00074943,MFCD00074909,MFCD20617670 | |
trans-4-methylcyclohexanecarboxylic acid, 4-methylcyclohexanecarboxylic acid, 4-methyl-1-cyclohexanecarboxylic acid, cis-4-methylcyclohexanecarboxylic acid, unii-70f0i2amtv, unii-m7dsk28350, trans-4-methyl-1-cyclohexanecarboxylic acid, trans-4-methyl hexahydrobenzoic acid, trans-4-methylcyclohexane carboxylic acid, 70f0i2amtv | |
CC1CCC(CC1)C(O)=O | |
142.20 | |
142.20 | |
Crystalline Powder |
13064-83-0 | |
White-Yellow | |
C8H14O2 | |
25 g | |
QTDXSEZXAPHVBI-UHFFFAOYSA-N | |
4-methylcyclohexane-1-carboxylic acid | |
20330 | |
≥98.0% (GC,T) |
Chemical Identifiers
13064-83-0 | |
142.20 | |
QTDXSEZXAPHVBI-UHFFFAOYSA-N | |
20330 | |
CC1CCC(CC1)C(O)=O |
C8H14O2 | |
MFCD00074943,MFCD00074909,MFCD20617670 | |
trans-4-methylcyclohexanecarboxylic acid, 4-methylcyclohexanecarboxylic acid, 4-methyl-1-cyclohexanecarboxylic acid, cis-4-methylcyclohexanecarboxylic acid, unii-70f0i2amtv, unii-m7dsk28350, trans-4-methyl-1-cyclohexanecarboxylic acid, trans-4-methyl hexahydrobenzoic acid, trans-4-methylcyclohexane carboxylic acid, 70f0i2amtv | |
4-methylcyclohexane-1-carboxylic acid |
Safety and Handling
EINECSNumber : (3)-3311
TSCA : No