missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-Methyl-beta-nitrostyrene, 98%

Manufacturer:  Alfa Aesar™ A1472114

 View more versions of this product

Catalog No. AAA1472114



Description

Description

Applications
trans-4-Methyl-β-nitrostyrene may be used as a reagent in the synthesis of N-benzylpyrrolomorphinans. It is also used as an organic intermediate for pharmaceutical.

Solubility
Sparingly Soluble in water (0.070 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

trans-4-Methyl-beta-nitrostyrene
25g
C9H9NO2
1864209
JSPNBERPFLONRX-VOTSOKGWSA-N
1-methyl-4-[(E)-2-nitroethenyl]benzene
674135
98%
100°C to 104°C
5153-68-4
MFCD00024828
1-methyl-4-2-nitrovinyl benzene, trans-4-methyl-beta-nitrostyrene, e-1-methyl-4-2-nitrovinyl benzene, 4-methyl-b-nitrostyrene, 4-methyl-beta-nitrostyrene, 1-methyl-4-2-nitroethenyl benzene, 1-methyl-4-e-2-nitroethenyl benzene, 4-methyl-.beta.-nitrostyrene, 4-methyl-.omega.-nitrostyrene, e-beta-nitro-4-methylstyrene
CC1=CC=C(C=C1)C=C[N+](=O)[O-]
163.176
163.18

Chemical Identifiers

5153-68-4
163.176
JSPNBERPFLONRX-VOTSOKGWSA-N
674135
CC1=CC=C(C=C1)C=C[N+](=O)[O-]
C9H9NO2
MFCD00024828
1-methyl-4-2-nitrovinyl benzene, trans-4-methyl-beta-nitrostyrene, e-1-methyl-4-2-nitrovinyl benzene, 4-methyl-b-nitrostyrene, 4-methyl-beta-nitrostyrene, 1-methyl-4-2-nitroethenyl benzene, 1-methyl-4-e-2-nitroethenyl benzene, 4-methyl-.beta.-nitrostyrene, 4-methyl-.omega.-nitrostyrene, e-beta-nitro-4-methylstyrene
1-methyl-4-[(E)-2-nitroethenyl]benzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

SDS Documents