missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-4-Methyl-beta-nitrostyrene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1472106

 View more versions of this product

Catalog No. AAA1472106



Description

Description

trans-4-Methyl-β-nitrostyrene may be used as a reagent in the synthesis of N-benzylpyrrolomorphinans. It is also used as an organic intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Methyl-β-nitrostyrene may be used as a reagent in the synthesis of N-benzylpyrrolomorphinans. It is also used as an organic intermediate for pharmaceutical.

Solubility
Sparingly Soluble in water (0.070 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

trans-4-Methyl-beta-nitrostyrene
100°C to 104°C
MFCD00024828
1864209
Sparingly Soluble in water (0.070g/L) (25°C).
CC1=CC=C(C=C1)C=C[N+](=O)[O-]
163.176
163.18
5153-68-4
C9H9NO2
5 g
1-methyl-4-2-nitrovinyl benzene, trans-4-methyl-beta-nitrostyrene, e-1-methyl-4-2-nitrovinyl benzene, 4-methyl-b-nitrostyrene, 4-methyl-beta-nitrostyrene, 1-methyl-4-2-nitroethenyl benzene, 1-methyl-4-e-2-nitroethenyl benzene, 4-methyl-.beta.-nitrostyrene, 4-methyl-.omega.-nitrostyrene, e-beta-nitro-4-methylstyrene
JSPNBERPFLONRX-VOTSOKGWSA-N
1-methyl-4-[(E)-2-nitroethenyl]benzene
674135
98%

Chemical Identifiers

5153-68-4
163.176
JSPNBERPFLONRX-VOTSOKGWSA-N
674135
CC1=CC=C(C=C1)C=C[N+](=O)[O-]
C9H9NO2
MFCD00024828
1-methyl-4-2-nitrovinyl benzene, trans-4-methyl-beta-nitrostyrene, e-1-methyl-4-2-nitrovinyl benzene, 4-methyl-b-nitrostyrene, 4-methyl-beta-nitrostyrene, 1-methyl-4-2-nitroethenyl benzene, 1-methyl-4-e-2-nitroethenyl benzene, 4-methyl-.beta.-nitrostyrene, 4-methyl-.omega.-nitrostyrene, e-beta-nitro-4-methylstyrene
1-methyl-4-[(E)-2-nitroethenyl]benzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only