missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-4-Methyl-2-pentene, TCI America™

Manufacturer:  TCI America M03945ML

 View more versions of this product

Catalog No. M03945ML



Specifications

Specifications

trans-4-Methyl-2-pentene
Colorless
674-76-0
MFCD00065138
LGAQJENWWYGFSN-SNAWJCMRSA-N
(E)-4-methylpent-2-ene
172092
Clear Liquid at 20°C
58°C
5mL
C6H12
2288
CC=CC(C)C
84.162
84.16
≥97.0% (GC)

Chemical Identifiers

674-76-0
84.162
LGAQJENWWYGFSN-SNAWJCMRSA-N
(E)-4-methylpent-2-ene
C6H12
MFCD00065138
172092
CC=CC(C)C
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0031

TSCA : No

Documents