missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-4-Methyl-1-cyclohexanecarboxylic acid, 98+%, ACROS Organics™

Chemical Identifiers

CAS 13064-83-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.2
MDL Number MFCD00074943
InChI Key QTDXSEZXAPHVBI-UHFFFAOYSA-N
Synonym 4-methyl-1-cyclohexanecarboxylic acid, 4-methylcyclohexanecarboxylic acid, 70f0i2amtv, cis-4-methylcyclohexanecarboxylic acid, trans-4-methyl hexahydrobenzoic acid, trans-4-methyl-1-cyclohexanecarboxylic acid, trans-4-methylcyclohexane carboxylic acid, trans-4-methylcyclohexanecarboxylic acid, unii-70f0i2amtv, unii-m7dsk28350
PubChem CID 20330
IUPAC Name 4-methylcyclohexane-1-carboxylic acid
SMILES CC1CCC(CC1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC427500010
SDS View Documents
Thermo Fisher Scientific
AC427500010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC427500050
SDS View Documents
Thermo Fisher Scientific
AC427500050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

13064-83-0
142.2
QTDXSEZXAPHVBI-UHFFFAOYSA-N
20330
CC1CCC(CC1)C(=O)O
C8H14O2
MFCD00074943
4-methyl-1-cyclohexanecarboxylic acid, 4-methylcyclohexanecarboxylic acid, 70f0i2amtv, cis-4-methylcyclohexanecarboxylic acid, trans-4-methyl hexahydrobenzoic acid, trans-4-methyl-1-cyclohexanecarboxylic acid, trans-4-methylcyclohexane carboxylic acid, trans-4-methylcyclohexanecarboxylic acid, unii-70f0i2amtv, unii-m7dsk28350
4-methylcyclohexane-1-carboxylic acid

Specifications

Glass bottle
109.0°C to 111.0°C
13064-83-0
C8H14O2
4-methyl-1-cyclohexanecarboxylic acid, 4-methylcyclohexanecarboxylic acid, 70f0i2amtv, cis-4-methylcyclohexanecarboxylic acid, trans-4-methyl hexahydrobenzoic acid, trans-4-methyl-1-cyclohexanecarboxylic acid, trans-4-methylcyclohexane carboxylic acid, trans-4-methylcyclohexanecarboxylic acid, unii-70f0i2amtv, unii-m7dsk28350
CC1CCC(CC1)C(=O)O
142.2
142.2
trans-4-Methyl-1-cyclohexanecarboxylic acid
245.0°C
1g
98+%
MFCD00074943
QTDXSEZXAPHVBI-UHFFFAOYSA-N
4-methylcyclohexane-1-carboxylic acid
20330
98+%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 235-959-6