Please login to your online account to display your discounted pricing

trans-4-Methoxycinnamaldehyde, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  35171-0250

 View more versions of this product

Catalog No. AC351710250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder or Chunks
Assay ≥97.5%
Formula Weight 162.19
Quantity 25g
Color Orange-yellow
Melting Point 55° to 60°C
Chemical Name or Material trans-4-Methoxycinnamaldehyde
MDL Number MFCD00017343
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Assay Percent Range 98%
Molecular Formula C10H10O2
Packaging Glass bottle
CAS 24680-50-0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material trans-4-Methoxycinnamaldehyde
CAS 24680-50-0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H10O2
Formula Weight 162.19
MDL Number MFCD00017343
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents