missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-4-Methoxycinnamaldehyde, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 35171-0050

 View more versions of this product

Catalog No. AC351710050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
162.19
Orange-yellow
Glass bottle
C10H10O2
MFCD00017343
Authentic
Crystalline Powder or Chunks
5g
55° to 60°C
24680-50-0
trans-4-Methoxycinnamaldehyde
98%

Chemical Identifiers

24680-50-0
C10H10O2
MFCD00017343
≥97.5%
98%
162.19
trans-4-Methoxycinnamaldehyde

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.