Please login to your online account to display your discounted pricing

trans-4-Hydroxycrotonic acid, Tocris


GHB receptor ligand

Quantity 10mg

Manufacturer: r & d systems  RDS053810

 View more versions of this product

Catalog No. 053810

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 102.09
Chemical Name or Material trans-4-Hydroxycrotonic acid
Assay Percent Range >99%
Molecular Formula C4H6O3
CAS 24587-49-3
Synonym trans-4-Hydroxy-2-butenoic acid
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material trans-4-Hydroxycrotonic acid
Assay Percent Range >99%
Molecular Formula C4H6O3
Formula Weight 102.09
Synonym trans-4-Hydroxy-2-butenoic acid
Accessories

Desiccate at +4°C