Please login to your online account to display your discounted pricing

trans-4-Hydroxycrotonic acid, Tocris

GHB receptor ligand

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material trans-4-Hydroxycrotonic acid
Assay Percent Range >99%
Molecular Formula C4H6O3
Formula Weight 102.09
Synonym trans-4-Hydroxy-2-butenoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

053810

 
r & d systems
RDS053810
10mg Each for $164.61

053850

 
r & d systems
RDS053850
50mg Each for $682.67
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 102.09
Molecular Formula C4H6O3
Chemical Name or Material trans-4-Hydroxycrotonic acid
Assay Percent Range >99%
CAS 24587-49-3
Synonym trans-4-Hydroxy-2-butenoic acid
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C