Learn More
Alfa Aesar™ trans-4-Hydroxycinnamic acid, 98%
Manufacturer: Alfa Aesar™ A1516722
Description
Applicationstrans-4-Hydroxycinnamic acid is used as a hydroxycinnamic acid with antioxidant properties. p-Coumaric acid is an aromatic phytochemical for proteomics research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
trans-4-Hydroxycinnamic acid | |
100g | |
C9H8O3 | |
2207383 | |
p-coumaric acid, 4-hydroxycinnamic acid, p-hydroxycinnamic acid, 4-coumaric acid, trans-4-hydroxycinnamic acid, trans-p-coumaric acid, p-cumaric acid, 3-4-hydroxyphenyl acrylic acid, para-coumaric acid, hydroxycinnamic acid | |
C1=CC(=CC=C1C=CC(=O)O)O | |
164.16 | |
CHEBI:32374 | |
98% |
~214°C (decomposition) | |
501-98-4 | |
MFCD00004399 | |
14,2560 | |
NGSWKAQJJWESNS-ZZXKWVIFSA-N | |
(E)-3-(4-hydroxyphenyl)prop-2-enoic acid | |
637542 | |
164.16 |
Chemical Identifiers
501-98-4 | |
164.16 | |
NGSWKAQJJWESNS-ZZXKWVIFSA-N | |
637542 | |
(E)-3-(4-hydroxyphenyl)prop-2-enoic acid |
C9H8O3 | |
MFCD00004399 | |
p-coumaric acid, 4-hydroxycinnamic acid, p-hydroxycinnamic acid, 4-coumaric acid, trans-4-hydroxycinnamic acid, trans-p-coumaric acid, p-cumaric acid, 3-4-hydroxyphenyl acrylic acid, para-coumaric acid, hydroxycinnamic acid | |
CHEBI:32374 | |
C1=CC(=CC=C1C=CC(=O)O)O |
Safety and Handling
GHS H Statement
H361-H315-H319-H335
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
RTECSNumber : GD9095000