Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-Hydroxy-L-proline, 99+%

$52.65 - $290.92

Chemical Identifiers

Chemical Name or Material trans-4-Hydroxy-L-proline
CAS 51-35-4
Molecular Formula C5H9NO3
Formula Weight 131.13
MDL Number MFCD00064320
Beilstein 471933
Merck Index 14,4840
Synonym (2S,4R)-(-)-4-Hydroxypyrrolidine-2-carboxylic acid; H-Hyp-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1185109

 
alfa aesar™
A1185109
10g Each for $52.65

AAA1185114

 
alfa aesar™
A1185114
25g Each for $99.74

AAA1185122

 
alfa aesar™
A1185122
100g Each for $290.92
Description & Specifications

Specifications

Formula Weight 131.13
Chemical Name or Material trans-4-Hydroxy-L-proline
TSCA TSCA
Assay ≥99%
Molecular Formula C5H9NO3
MDL Number MFCD00064320
CAS 51-35-4
Beilstein 471933
Merck Index 14,4840
Synonym (2S,4R)-(-)-4-Hydroxypyrrolidine-2-carboxylic acid; H-Hyp-OH
Melting Point 270° to 273°C (dec.)
Quantity 10g
Optical Rotation −76° (c=4 in water)
RTECS Number TW3586500

Applications
trans-4-Hydroxy-L-proline is used as a reagent for the preparation of neuroexcitatory kainoids and antifungal echinocandins. It is also involved in the synthesis of chiral ligands for enantioselective ethylation of aldehydes.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.