missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-Hydroxy-L-proline, 99+%

Manufacturer:  Alfa Aesar™ A1185114

 View more versions of this product

Catalog No. AAA1185114



Description

Description

Applications
trans-4-Hydroxy-L-proline is used as a reagent for the preparation of neuroexcitatory kainoids and antifungal echinocandins. It is also involved in the synthesis of chiral ligands for enantioselective ethylation of aldehydes.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

trans-4-Hydroxy-L-proline
≥99%
MFCD00064320
14,4840
PMMYEEVYMWASQN-DMTCNVIQSA-N
(2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
5810
131.13
25g
51-35-4
C5H9NO3
471933
(2S,4R)-(-)-4-Hydroxypyrrolidine-2-carboxylic acid; H-Hyp-OH
C1C(CNC1C(=O)O)O
131.131
CHEBI:18095
270°C to 273°C (decomposition)
−76° (c=4 in water)

Chemical Identifiers

51-35-4
131.131
PMMYEEVYMWASQN-DMTCNVIQSA-N
5810
(2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
C5H9NO3
MFCD00064320
(2S,4R)-(-)-4-Hydroxypyrrolidine-2-carboxylic acid; H-Hyp-OH
CHEBI:18095
C1C(CNC1C(=O)O)O
Safety and Handling

Safety and Handling

RTECSNumber : TW3586500

TSCA : TSCA

SDS Documents