missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals trans-4-Hydroxy-L-proline, 99+%

Catalog No. AC121780100
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC121780100 10 g Glass bottle
AC121780250 25 g Glass bottle
AC121781000 100 g Glass bottle
AC121785000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC121780100 Supplier Thermo Scientific Chemicals Supplier No. 121780100
Only null left

CAS: 51-35-4 | C5H9NO3 | 131.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51-35-4
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
InChI Key PMMYEEVYMWASQN-DMTCNVIQSA-N
Synonym trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro
PubChem CID 5810
ChEBI CHEBI:18095
IUPAC Name (2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES C1C(CNC1C(=O)O)O

Specifications

Melting Point 274.0°C to 275.0°C
Color White
Loss on Drying 0.2% (105°C, 3 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 10 g
Beilstein 22,191
Merck Index 15,4878
Specific Rotation Condition - 75.50 (20.00°C c=5,H2O)
Specific Rotation − 75.50
Solubility Information Solubility in water: 357.8g/L (20° c). Other solubilities: slightly soluble in alcohol, insoluble in ether
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Formula Weight 131.13
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material trans-4-Hydroxy-L-proline
Show More Show Less
EINECSNumber 200-091-9

RUO – Research Use Only