Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-Hydroxy-1-(4-nitrobenzyloxycarbonyl)-L-proline, 97%

Chemical Identifiers

Chemical Name or Material trans-4-Hydroxy-1-(4-nitrobenzyloxycarbonyl)-L-proline
CAS 96034-57-0
Formula Weight 310.26
Synonym (2S,4R)-1-((4-nitrobenzyloxy)carbonyl)-4-hydroxypyrrolidine-2-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6119403

 
alfa aesar™
H6119403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6119406

 
alfa aesar™
H6119406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 310.26
Chemical Name or Material trans-4-Hydroxy-1-(4-nitrobenzyloxycarbonyl)-L-proline
Physical Form Solid
Assay 97%
CAS 96034-57-0
Synonym (2S,4R)-1-((4-nitrobenzyloxy)carbonyl)-4-hydroxypyrrolidine-2-carboxylic acid
Quantity 1g