Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-Ethylcyclohexanecarboxylic acid, 98%

$220.12 - $220.12

Chemical Identifiers

Chemical Name or Material trans-4-Ethylcyclohexanecarboxylic acid
CAS 6833-47-2
Molecular Formula C9H16O2
Formula Weight 156.23
MDL Number MFCD01076323
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2189706

 
alfa aesar™
B2189706
5g Each for $220.12
Description & Specifications

Specifications

Formula Weight 156.23
Chemical Name or Material trans-4-Ethylcyclohexanecarboxylic acid
Assay 98%
Molecular Formula C9H16O2
MDL Number MFCD01076323
CAS 6833-47-2
Melting Point 48° to 51°C
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.