missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-Ethyl-4'-(4-n-pentylcyclohexyl)biphenyl, 97%

$79.04 - $314.09

Chemical Identifiers

CAS 79709-85-6
Molecular Formula C25H34
Molecular Weight (g/mol) 334.547
MDL Number MFCD01941072
InChI Key JOLGXBQYTARJLD-UHFFFAOYSA-N
Synonym 1,1'-biphenyl, 4-ethyl-4'-4-pentylcyclohexyl
PubChem CID 630553
IUPAC Name 1-ethyl-4-[4-(4-pentylcyclohexyl)phenyl]benzene
SMILES CCCCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC=C(C=C3)CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5564703 View Documents Alfa Aesar™
H5564703
1g Each for $79.04
 
AAH5564706 View Documents Alfa Aesar™
H5564706
5g Each for $314.09
 
Specifications

Chemical Identifiers

79709-85-6
334.547
JOLGXBQYTARJLD-UHFFFAOYSA-N
630553
CCCCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC=C(C=C3)CC
C25H34
MFCD01941072
1,1'-biphenyl, 4-ethyl-4'-4-pentylcyclohexyl
1-ethyl-4-[4-(4-pentylcyclohexyl)phenyl]benzene

Specifications

145°C to 147°C
79709-85-6
MFCD01941072
JOLGXBQYTARJLD-UHFFFAOYSA-N
1-ethyl-4-[4-(4-pentylcyclohexyl)phenyl]benzene
630553
97%
1g
C25H34
1,1'-biphenyl, 4-ethyl-4'-4-pentylcyclohexyl
CCCCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC=C(C=C3)CC
334.547
334.54
trans-4-Ethyl-4'-(4-n-pentylcyclohexyl)biphenyl
Safety and Handling

Safety and Handling

EINECSNumber : 279-240-5

TSCA : No

Recommended Storage : Ambient temperatures