missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-Ethyl-4'-(4-n-pentylcyclohexyl)biphenyl, 97%

Manufacturer:  Alfa Aesar™ H5564703

 View more versions of this product

Catalog No. AAH5564703



Specifications

Specifications

trans-4-Ethyl-4'-(4-n-pentylcyclohexyl)biphenyl
1g
C25H34
1,1'-biphenyl, 4-ethyl-4'-4-pentylcyclohexyl
CCCCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC=C(C=C3)CC
334.547
334.54
145°C to 147°C
79709-85-6
MFCD01941072
JOLGXBQYTARJLD-UHFFFAOYSA-N
1-ethyl-4-[4-(4-pentylcyclohexyl)phenyl]benzene
630553
97%

Chemical Identifiers

79709-85-6
334.547
JOLGXBQYTARJLD-UHFFFAOYSA-N
630553
CCCCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC=C(C=C3)CC
C25H34
MFCD01941072
1,1'-biphenyl, 4-ethyl-4'-4-pentylcyclohexyl
1-ethyl-4-[4-(4-pentylcyclohexyl)phenyl]benzene
Safety and Handling

Safety and Handling

EINECSNumber : 279-240-5

TSCA : No

Recommended Storage : Ambient temperatures

Documents