missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Cyanocyclohexylamine hydrochloride, 97%

CAS: 873537-33-8 | C7H13ClN2 | 160.645 g/mol

Supplier:  Thermo Scientific Chemicals H62522MD

 View more versions of this product

Catalog No. AAH62522MD


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in a cool and dry condition. Incompatible with oxidizing agents.
Specifications

Specifications

trans-4-Cyanocyclohexylamine hydrochloride
C7H13ClN2
250 mg
trans-4-aminocyclohexanecarbonitrile hydrochloride, cis-4-aminocyclohexanecarbonitrile hydrochloride, 4-amino-cyclohexanecarbonitrile hydrochloride, 4-aminocyclohexanecarbonitrile hydrochloride, 4-aminocyclohexane-1-carbonitrile hydrochloride, c7h12n2.hcl, 1s,4s-4-aminocyclohexanecarbonitrile hydrochloride, 1r,4r-4-aminocyclohexane-1-carbonitrile hydrochloride, 1s,4s-4-aminocyclohexane-1-carbonitrile hydrochloride, cyclohexanecarbonitrile, 4-amino-, monohydrochloride, trans
C1CC(CCC1C#N)N.Cl
160.645
160.64
873537-33-8
MFCD21643021
UN3439
BTLVSLNZSMIPCW-UHFFFAOYSA-N
4-aminocyclohexane-1-carbonitrile;hydrochloride
53407876
97%

Chemical Identifiers

873537-33-8
160.645
BTLVSLNZSMIPCW-UHFFFAOYSA-N
53407876
C1CC(CCC1C#N)N.Cl
C7H13ClN2
MFCD21643021
trans-4-aminocyclohexanecarbonitrile hydrochloride, cis-4-aminocyclohexanecarbonitrile hydrochloride, 4-amino-cyclohexanecarbonitrile hydrochloride, 4-aminocyclohexanecarbonitrile hydrochloride, 4-aminocyclohexane-1-carbonitrile hydrochloride, c7h12n2.hcl, 1s,4s-4-aminocyclohexanecarbonitrile hydrochloride, 1r,4r-4-aminocyclohexane-1-carbonitrile hydrochloride, 1s,4s-4-aminocyclohexane-1-carbonitrile hydrochloride, cyclohexanecarbonitrile, 4-amino-, monohydrochloride, trans
4-aminocyclohexane-1-carbonitrile;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only