missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-(Boc-aminomethyl)cyclohexanemethylamine, 97%

$43.81 - $518.15

Chemical Identifiers

CAS 166168-16-7
Molecular Formula C13H26N2O2
Molecular Weight (g/mol) 242.363
MDL Number MFCD04112680
InChI Key NYXOBVVHJZENCO-UHFFFAOYSA-N
Synonym tert-butyl trans-4-aminomethyl cyclohexyl methyl carbamate, trans-l-4-boc-aminomethyl-cyclohexane methanamine, trans-l-4-boc-aminomethyl cyclohexane methanamine, trans-4-aminomethyl-cyclohexylmethyl-carbamic acid tert-butyl ester, trans-4-boc-aminomethyl cyclohexanemethanamine, tert-butyl n-4-aminomethyl cyclohexyl methyl carbamate, tert-butyl n-4-aminomethyl cyclohexyl methyl carbamate, n-t-butyloxycarbonyl-1,4-trans-diaminomethyl-cyclohexane, carbamicacid, n-trans-4-aminomethyl cyclohexyl methyl-, 1,1-dimethylethyl ester, tert-butyl trans-4-aminomethyl cyclohexyl methyl carbamate
PubChem CID 2756046
IUPAC Name tert-butyl N-[[4-(aminomethyl)cyclohexyl]methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1CCC(CC1)CN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62906MD View Documents Alfa Aesar™
H62906MD
250mg Each for $43.81
 
AAH6290603 View Documents Alfa Aesar™
H6290603
1g Each for $130.00
 
AAH6290606 View Documents Alfa Aesar™
H6290606
5g Each for $518.15
 
Description

Description

Applications
Used as pharmaecutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

166168-16-7
242.363
NYXOBVVHJZENCO-UHFFFAOYSA-N
2756046
CC(C)(C)OC(=O)NCC1CCC(CC1)CN
C13H26N2O2
MFCD04112680
tert-butyl trans-4-aminomethyl cyclohexyl methyl carbamate, trans-l-4-boc-aminomethyl-cyclohexane methanamine, trans-l-4-boc-aminomethyl cyclohexane methanamine, trans-4-aminomethyl-cyclohexylmethyl-carbamic acid tert-butyl ester, trans-4-boc-aminomethyl cyclohexanemethanamine, tert-butyl n-4-aminomethyl cyclohexyl methyl carbamate, tert-butyl n-4-aminomethyl cyclohexyl methyl carbamate, n-t-butyloxycarbonyl-1,4-trans-diaminomethyl-cyclohexane, carbamicacid, n-trans-4-aminomethyl cyclohexyl methyl-, 1,1-dimethylethyl ester, tert-butyl trans-4-aminomethyl cyclohexyl methyl carbamate
tert-butyl N-[[4-(aminomethyl)cyclohexyl]methyl]carbamate

Specifications

250mg
C13H26N2O2
tert-butyl trans-4-aminomethyl cyclohexyl methyl carbamate, trans-l-4-boc-aminomethyl-cyclohexane methanamine, trans-l-4-boc-aminomethyl cyclohexane methanamine, trans-4-aminomethyl-cyclohexylmethyl-carbamic acid tert-butyl ester, trans-4-boc-aminomethyl cyclohexanemethanamine, tert-butyl n-4-aminomethyl cyclohexyl methyl carbamate, tert-butyl n-4-aminomethyl cyclohexyl methyl carbamate, n-t-butyloxycarbonyl-1,4-trans-diaminomethyl-cyclohexane, carbamicacid, n-trans-4-aminomethyl cyclohexyl methyl-, 1,1-dimethylethyl ester, tert-butyl trans-4-aminomethyl cyclohexyl methyl carbamate
CC(C)(C)OC(=O)NCC1CCC(CC1)CN
242.363
242.35
trans-4-(Boc-aminomethyl)cyclohexanemethylamine
166168-16-7
MFCD04112680
NYXOBVVHJZENCO-UHFFFAOYSA-N
tert-butyl N-[[4-(aminomethyl)cyclohexyl]methyl]carbamate
2756046
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning