missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-4-(Boc-aminomethyl)cyclohexanemethylamine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6290603

 View more versions of this product

Catalog No. AAH6290603



Description

Description

Used as pharmaecutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaecutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

trans-4-(Boc-aminomethyl)cyclohexanemethylamine
C13H26N2O2
tert-butyl trans-4-aminomethyl cyclohexyl methyl carbamate, trans-l-4-boc-aminomethyl-cyclohexane methanamine, trans-l-4-boc-aminomethyl cyclohexane methanamine, trans-4-aminomethyl-cyclohexylmethyl-carbamic acid tert-butyl ester, trans-4-boc-aminomethyl cyclohexanemethanamine, tert-butyl n-4-aminomethyl cyclohexyl methyl carbamate, n-t-butyloxycarbonyl-1,4-trans-diaminomethyl-cyclohexane, carbamicacid, n-trans-4-aminomethyl cyclohexyl methyl-, 1,1-dimethylethyl ester
NYXOBVVHJZENCO-UHFFFAOYSA-N
tert-butyl N-[[4-(aminomethyl)cyclohexyl]methyl]carbamate
2756046
97%
166168-16-7
MFCD04112680
Slightly soluble in water.
CC(C)(C)OC(=O)NCC1CCC(CC1)CN
242.363
242.35
1 g

Chemical Identifiers

166168-16-7
242.363
NYXOBVVHJZENCO-UHFFFAOYSA-N
2756046
CC(C)(C)OC(=O)NCC1CCC(CC1)CN
C13H26N2O2
MFCD04112680
tert-butyl trans-4-aminomethyl cyclohexyl methyl carbamate, trans-l-4-boc-aminomethyl-cyclohexane methanamine, trans-l-4-boc-aminomethyl cyclohexane methanamine, trans-4-aminomethyl-cyclohexylmethyl-carbamic acid tert-butyl ester, trans-4-boc-aminomethyl cyclohexanemethanamine, tert-butyl n-4-aminomethyl cyclohexyl methyl carbamate, n-t-butyloxycarbonyl-1,4-trans-diaminomethyl-cyclohexane, carbamicacid, n-trans-4-aminomethyl cyclohexyl methyl-, 1,1-dimethylethyl ester
tert-butyl N-[[4-(aminomethyl)cyclohexyl]methyl]carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only