missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-(Boc-aminomethyl)cyclohexanemethanol, 97%

$59.69 - $718.62

Chemical Identifiers

CAS 172348-63-9
Molecular Formula C13H25NO3
Molecular Weight (g/mol) 243.347
MDL Number MFCD06657666
InChI Key KSXPGASLEFGROT-UHFFFAOYSA-N
Synonym tert-butyl trans-4-hydroxymethylcyclohexylmethyl-carbamate, trans-4-boc-aminomethyl-1-hydroxymethyl-cyclohexane, tert-butyl trans-4-hydroxymethyl cyclohexyl methyl carbamate, tert-butyl trans-4-hydroxymethylcyclohexylmethyl carbamate, tert-butyl 4-hydroxymethyl cyclohexyl methylcarbamate, ksxpgaslefgrot-xypyzodxsa-n, tert-butyl trans-4-hydroxymethylcyclohexylmethyl carbamate, tert-butyl 4-hydroxymethyl cyclohexyl methyl carbamate, trans-4-n-tert-butoxycarbonylaminomethyl cyclohexylmethanol, n-tert-butoxycarbonyl-trans-4-aminomethyl cyclohexanemethanol
PubChem CID 18397396
IUPAC Name tert-butyl N-[[4-(hydroxymethyl)cyclohexyl]methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1CCC(CC1)CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62798MD View Documents Alfa Aesar™
H62798MD
250mg Each for $59.69
 
AAH6279803 View Documents Alfa Aesar™
H6279803
1g Each for $178.89
 
AAH6279806 View Documents Alfa Aesar™
H6279806
5g Each for $718.62
 
Specifications

Chemical Identifiers

172348-63-9
243.347
KSXPGASLEFGROT-UHFFFAOYSA-N
18397396
CC(C)(C)OC(=O)NCC1CCC(CC1)CO
C13H25NO3
MFCD06657666
tert-butyl trans-4-hydroxymethylcyclohexylmethyl-carbamate, trans-4-boc-aminomethyl-1-hydroxymethyl-cyclohexane, tert-butyl trans-4-hydroxymethyl cyclohexyl methyl carbamate, tert-butyl trans-4-hydroxymethylcyclohexylmethyl carbamate, tert-butyl 4-hydroxymethyl cyclohexyl methylcarbamate, ksxpgaslefgrot-xypyzodxsa-n, tert-butyl trans-4-hydroxymethylcyclohexylmethyl carbamate, tert-butyl 4-hydroxymethyl cyclohexyl methyl carbamate, trans-4-n-tert-butoxycarbonylaminomethyl cyclohexylmethanol, n-tert-butoxycarbonyl-trans-4-aminomethyl cyclohexanemethanol
tert-butyl N-[[4-(hydroxymethyl)cyclohexyl]methyl]carbamate

Specifications

250mg
C13H25NO3
tert-butyl trans-4-hydroxymethylcyclohexylmethyl-carbamate, trans-4-boc-aminomethyl-1-hydroxymethyl-cyclohexane, tert-butyl trans-4-hydroxymethyl cyclohexyl methyl carbamate, tert-butyl trans-4-hydroxymethylcyclohexylmethyl carbamate, tert-butyl 4-hydroxymethyl cyclohexyl methylcarbamate, ksxpgaslefgrot-xypyzodxsa-n, tert-butyl trans-4-hydroxymethylcyclohexylmethyl carbamate, tert-butyl 4-hydroxymethyl cyclohexyl methyl carbamate, trans-4-n-tert-butoxycarbonylaminomethyl cyclohexylmethanol, n-tert-butoxycarbonyl-trans-4-aminomethyl cyclohexanemethanol
CC(C)(C)OC(=O)NCC1CCC(CC1)CO
243.347
243.34
trans-4-(Boc-aminomethyl)cyclohexanemethanol
172348-63-9
MFCD06657666
KSXPGASLEFGROT-UHFFFAOYSA-N
tert-butyl N-[[4-(hydroxymethyl)cyclohexyl]methyl]carbamate
18397396
97%