missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-(Boc-amino)cyclohexanemethylamine, 97%

$79.11 - $1,047.09

Chemical Identifiers

CAS 177583-27-6
Molecular Formula C12H24N2O2
Molecular Weight (g/mol) 228.336
MDL Number MFCD01861810
InChI Key NVQFOBONHIXDOC-UHFFFAOYSA-N
Synonym tert-butyl trans-4-aminomethyl cyclohexyl carbamate, tert-butyl n-4-aminomethyl cyclohexyl carbamate, tert-butyl cis-4-aminomethylcyclohexylcarbamate, tert-butyl n-1r,4r-4-aminomethyl cyclohexyl carbamate, tert-butyl trans-4-aminomethylcyclohexyl carbamate, tert-butyl cis-4-aminomethyl cyclohexyl carbamate, cis-4-boc-amino cyclohexylmethylamine, tert-butyl trans-4-aminomethylcyclohexylcarbamate, trans-4-boc-amino cyclohexylmethylamine, tert-butyl 4-aminomethyl cyclohexyl carbamate
PubChem CID 2756045
IUPAC Name tert-butyl N-[4-(aminomethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62062MD View Documents Alfa Aesar™
H62062MD
250mg Each for $79.11
 
AAH6206203 View Documents Alfa Aesar™
H6206203
1g Each for $251.36
 
AAH6206206 View Documents Alfa Aesar™
H6206206
5g Each for $1,047.09
 
Description

Description

Applications
trans-4-(Boc-amino)cyclohexanemethylamine is used as a raw material intermediates for pharmaceutical and chemical.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

177583-27-6
228.336
NVQFOBONHIXDOC-UHFFFAOYSA-N
2756045
CC(C)(C)OC(=O)NC1CCC(CC1)CN
C12H24N2O2
MFCD01861810
tert-butyl trans-4-aminomethyl cyclohexyl carbamate, tert-butyl n-4-aminomethyl cyclohexyl carbamate, tert-butyl cis-4-aminomethylcyclohexylcarbamate, tert-butyl n-1r,4r-4-aminomethyl cyclohexyl carbamate, tert-butyl trans-4-aminomethylcyclohexyl carbamate, tert-butyl cis-4-aminomethyl cyclohexyl carbamate, cis-4-boc-amino cyclohexylmethylamine, tert-butyl trans-4-aminomethylcyclohexylcarbamate, trans-4-boc-amino cyclohexylmethylamine, tert-butyl 4-aminomethyl cyclohexyl carbamate
tert-butyl N-[4-(aminomethyl)cyclohexyl]carbamate

Specifications

250mg
C12H24N2O2
tert-butyl trans-4-aminomethyl cyclohexyl carbamate, tert-butyl n-4-aminomethyl cyclohexyl carbamate, tert-butyl cis-4-aminomethylcyclohexylcarbamate, tert-butyl n-1r,4r-4-aminomethyl cyclohexyl carbamate, tert-butyl trans-4-aminomethylcyclohexyl carbamate, tert-butyl cis-4-aminomethyl cyclohexyl carbamate, cis-4-boc-amino cyclohexylmethylamine, tert-butyl trans-4-aminomethylcyclohexylcarbamate, trans-4-boc-amino cyclohexylmethylamine, tert-butyl 4-aminomethyl cyclohexyl carbamate
CC(C)(C)OC(=O)NC1CCC(CC1)CN
228.336
228.33
trans-4-(Boc-amino)cyclohexanemethylamine
177583-27-6
MFCD01861810
NVQFOBONHIXDOC-UHFFFAOYSA-N
tert-butyl N-[4-(aminomethyl)cyclohexyl]carbamate
2756045
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning