Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-(Boc-amino)cyclohexanecarboxylic acid, 98%

Chemical Identifiers

Chemical Name or Material trans-4-(Boc-amino)cyclohexanecarboxylic acid
CAS 53292-89-0
Formula Weight 243.3
MDL Number MFCD01862293
Beilstein 7641071
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5940201

 
alfa aesar™
H5940201
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH59402ME

 
alfa aesar™
H59402ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 243.3
Chemical Name or Material trans-4-(Boc-amino)cyclohexanecarboxylic acid
Physical Form Powder
Assay 98%
MDL Number MFCD01862293
CAS 53292-89-0
Beilstein 7641071
Color White
Quantity 0.25g