missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-(Boc-amino)cyclohexaneacetaldehyde, 97%

$142.03 - $465.72

Chemical Identifiers

CAS 215790-29-7
Molecular Formula C13H23NO3
Molecular Weight (g/mol) 241.331
MDL Number MFCD03844592
InChI Key OPGBSEXLSWYFOR-UHFFFAOYSA-N
Synonym tert-butyl trans-4-2-oxoethyl cyclohexylcarbamate, tert-butyl cis-4-2-oxoethyl cyclohexylcarbamate, tert-butyl n-4-2-oxoethyl cyclohexyl carbamate, carbamic acid,n-trans-4-2-oxoethyl cyclohexyl-, 1,1-dimethylethyl ester, opgbsexlswyfor-xypyzodxsa-n, tert-butyl 4-2-oxoethyl cyclohexyl carbamate, cis-tert-butyl 4-2-oxoethyl cyclohexyl carbamate, trans-tert-butyl-4-2-oxoethyl-cyclohexylcarbamate, cis-4-tert-butoxycarbonylamino cyclohexylacetaldehyde, tert-butyl trans-4-2-oxoethyl cyclohexyl carbamate
PubChem CID 24720884
IUPAC Name tert-butyl N-[4-(2-oxoethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CC=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62373MD View Documents Alfa Aesar™
H62373MD
250mg Each for $142.03
 
AAH6237303 View Documents Alfa Aesar™
H6237303
1g Each for $465.72
 
Description

Description

Applications
trans-4-(Boc-amino)cyclohexaneacetaldehyde is an intermediate for the synthesis of cariprazine (C183490), a novel antipsychotic acting on dopamine D3/D2 receptors.

Solubility
Sparingly Soluble (0.41 g/L) (25°C).

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Chemical Identifiers

215790-29-7
241.331
OPGBSEXLSWYFOR-UHFFFAOYSA-N
24720884
CC(C)(C)OC(=O)NC1CCC(CC1)CC=O
C13H23NO3
MFCD03844592
tert-butyl trans-4-2-oxoethyl cyclohexylcarbamate, tert-butyl cis-4-2-oxoethyl cyclohexylcarbamate, tert-butyl n-4-2-oxoethyl cyclohexyl carbamate, carbamic acid,n-trans-4-2-oxoethyl cyclohexyl-, 1,1-dimethylethyl ester, opgbsexlswyfor-xypyzodxsa-n, tert-butyl 4-2-oxoethyl cyclohexyl carbamate, cis-tert-butyl 4-2-oxoethyl cyclohexyl carbamate, trans-tert-butyl-4-2-oxoethyl-cyclohexylcarbamate, cis-4-tert-butoxycarbonylamino cyclohexylacetaldehyde, tert-butyl trans-4-2-oxoethyl cyclohexyl carbamate
tert-butyl N-[4-(2-oxoethyl)cyclohexyl]carbamate

Specifications

250mg
C13H23NO3
tert-butyl trans-4-2-oxoethyl cyclohexylcarbamate, tert-butyl cis-4-2-oxoethyl cyclohexylcarbamate, tert-butyl n-4-2-oxoethyl cyclohexyl carbamate, carbamic acid,n-trans-4-2-oxoethyl cyclohexyl-, 1,1-dimethylethyl ester, opgbsexlswyfor-xypyzodxsa-n, tert-butyl 4-2-oxoethyl cyclohexyl carbamate, cis-tert-butyl 4-2-oxoethyl cyclohexyl carbamate, trans-tert-butyl-4-2-oxoethyl-cyclohexylcarbamate, cis-4-tert-butoxycarbonylamino cyclohexylacetaldehyde, tert-butyl trans-4-2-oxoethyl cyclohexyl carbamate
CC(C)(C)OC(=O)NC1CCC(CC1)CC=O
241.331
241.32
trans-4-(Boc-amino)cyclohexaneacetaldehyde
215790-29-7
MFCD03844592
OPGBSEXLSWYFOR-UHFFFAOYSA-N
tert-butyl N-[4-(2-oxoethyl)cyclohexyl]carbamate
24720884
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning