Learn More
trans-4-(Benzyloxycarbonylamino)cyclohexylamine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H62444MD
Description
trans-4-(Benzyloxycarbonylamino)cyclohexylamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans-4-(Benzyloxycarbonylamino)cyclohexylamine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
trans-4-(Benzyloxycarbonylamino)cyclohexylamine | |
C14H20N2O2 | |
250 mg | |
Slightly soluble in water. | |
NC1CCC(CC1)NC(=O)OCC1=CC=CC=C1 | |
248.33 | |
248.32 |
149423-77-8 | |
MFCD03839901,MFCD06657668,MFCD07772066 | |
benzyl cis-4-aminocyclohexyl carbamate, benzyl trans-4-aminocyclohexyl carbamate, n-cbz-trans-1,4-cyclohexanediamine, cis-1-n-cbz-1,4-cyclohexyldiamine, cis-n-cbz-1,4-cyclohexanediamine, 4-amino-cyclohexyl-carbamic acid benzyl ester, benzyl cis-4-aminocyclohexylcarbamate, benzyl 4-aminocyclohexyl carbamate, benzyl 1r,4r-4-aminocyclohexyl carbamate, 1-n-cbz-trans-1,4-cyclohexyldiamine | |
JQVBZZUMWRXDSQ-UHFFFAOYSA-N | |
benzyl N-(4-aminocyclohexyl)carbamate | |
18650082 | |
97% |
Chemical Identifiers
149423-77-8 | |
248.33 | |
JQVBZZUMWRXDSQ-UHFFFAOYSA-N | |
18650082 | |
NC1CCC(CC1)NC(=O)OCC1=CC=CC=C1 |
C14H20N2O2 | |
MFCD03839901,MFCD06657668,MFCD07772066 | |
benzyl cis-4-aminocyclohexyl carbamate, benzyl trans-4-aminocyclohexyl carbamate, n-cbz-trans-1,4-cyclohexanediamine, cis-1-n-cbz-1,4-cyclohexyldiamine, cis-n-cbz-1,4-cyclohexanediamine, 4-amino-cyclohexyl-carbamic acid benzyl ester, benzyl cis-4-aminocyclohexylcarbamate, benzyl 4-aminocyclohexyl carbamate, benzyl 1r,4r-4-aminocyclohexyl carbamate, 1-n-cbz-trans-1,4-cyclohexyldiamine | |
benzyl N-(4-aminocyclohexyl)carbamate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only