missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine hydrochloride, 97%

$70.62 - $1,025.51

Chemical Identifiers

CAS 187533-10-4
Molecular Formula C15H23ClN2O2
Molecular Weight (g/mol) 298.811
InChI Key RUVDEZPQPDTJBK-UHFFFAOYSA-N
Synonym ruvdezpqpdtjbk-jgoftkabsa-n, benzyl n-4-aminomethyl cyclohexyl carbamate hcl, benzyl trans-4-aminomethylcyclohexylcarbamate hydrochloride, benzyl trans-4-aminomethylcyclohexyl-carbamate hydrochloride, benzyl n-1r,4r-4-aminomethyl cyclohexyl carbamate hydrochloride, trans-4-aminomethyl-cyclohexyl-carbamic acid benzyl ester hydrochloride, trans-1-benzyloxycarbonyl amino-4-aminomethylcyclohexane hydrochloride
PubChem CID 22990186
IUPAC Name benzyl N-[4-(aminomethyl)cyclohexyl]carbamate;hydrochloride
SMILES C1CC(CCC1CN)NC(=O)OCC2=CC=CC=C2.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62693MD View Documents Alfa Aesar™
H62693MD
250mg Each for $70.62
 
AAH6269303 View Documents Alfa Aesar™
H6269303
1g Each for $225.15
 
AAH6269306 View Documents Alfa Aesar™
H6269306
5g Each for $1,025.51
 
Specifications

Chemical Identifiers

187533-10-4
298.811
ruvdezpqpdtjbk-jgoftkabsa-n, benzyl n-4-aminomethyl cyclohexyl carbamate hcl, benzyl trans-4-aminomethylcyclohexylcarbamate hydrochloride, benzyl trans-4-aminomethylcyclohexyl-carbamate hydrochloride, benzyl n-1r,4r-4-aminomethyl cyclohexyl carbamate hydrochloride, trans-4-aminomethyl-cyclohexyl-carbamic acid benzyl ester hydrochloride, trans-1-benzyloxycarbonyl amino-4-aminomethylcyclohexane hydrochloride
benzyl N-[4-(aminomethyl)cyclohexyl]carbamate;hydrochloride
C15H23ClN2O2
RUVDEZPQPDTJBK-UHFFFAOYSA-N
22990186
C1CC(CCC1CN)NC(=O)OCC2=CC=CC=C2.Cl

Specifications

250mg
C15H23ClN2O2
RUVDEZPQPDTJBK-UHFFFAOYSA-N
benzyl N-[4-(aminomethyl)cyclohexyl]carbamate;hydrochloride
22990186
97%
187533-10-4
ruvdezpqpdtjbk-jgoftkabsa-n, benzyl n-4-aminomethyl cyclohexyl carbamate hcl, benzyl trans-4-aminomethylcyclohexylcarbamate hydrochloride, benzyl trans-4-aminomethylcyclohexyl-carbamate hydrochloride, benzyl n-1r,4r-4-aminomethyl cyclohexyl carbamate hydrochloride, trans-4-aminomethyl-cyclohexyl-carbamic acid benzyl ester hydrochloride, trans-1-benzyloxycarbonyl amino-4-aminomethylcyclohexane hydrochloride
C1CC(CCC1CN)NC(=O)OCC2=CC=CC=C2.Cl
298.811
298.8
trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning