missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid, 97%

$28.84 - $337.73

Chemical Identifiers

CAS 34771-04-5
Molecular Formula C15H19NO4
Molecular Weight (g/mol) 277.32
MDL Number MFCD01550737
InChI Key ZVMICQYOGWAOSU-UHFFFAOYSA-N
Synonym trans-4-carbobenzoxyamino cyclohexanecarboxylic acid, trans-4-cbz-amino cyclohexanecarboxylic acid, 4-benzyloxycarbonylamino-cyclohexanecarboxylic acid, trans-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexane-carboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-phenylmethoxycarbonylamino cyclohexane-1-carboxylic acid, trans-4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid
PubChem CID 285247
IUPAC Name 4-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)NC(=O)OCC2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62926MD View Documents Alfa Aesar™
H62926MD
250mg Each for $28.84
 
AAH6292603 View Documents Alfa Aesar™
H6292603
1g Each for $85.14
 
AAH6292606 View Documents Alfa Aesar™
H6292606
5g Each for $337.73
 
Specifications

Chemical Identifiers

34771-04-5
277.32
ZVMICQYOGWAOSU-UHFFFAOYSA-N
285247
C1CC(CCC1C(=O)O)NC(=O)OCC2=CC=CC=C2
C15H19NO4
MFCD01550737
trans-4-carbobenzoxyamino cyclohexanecarboxylic acid, trans-4-cbz-amino cyclohexanecarboxylic acid, 4-benzyloxycarbonylamino-cyclohexanecarboxylic acid, trans-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexane-carboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-phenylmethoxycarbonylamino cyclohexane-1-carboxylic acid, trans-4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid
4-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid

Specifications

217°C
34771-04-5
MFCD01550737
ZVMICQYOGWAOSU-UHFFFAOYSA-N
4-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
285247
97%
250mg
C15H19NO4
trans-4-carbobenzoxyamino cyclohexanecarboxylic acid, trans-4-cbz-amino cyclohexanecarboxylic acid, 4-benzyloxycarbonylamino-cyclohexanecarboxylic acid, trans-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexane-carboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-phenylmethoxycarbonylamino cyclohexane-1-carboxylic acid, trans-4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid
C1CC(CCC1C(=O)O)NC(=O)OCC2=CC=CC=C2
277.32
277.31
trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid