missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-Benzyloxy-N-Fmoc-L-proline, 98%

$40.41 - $527.41

Chemical Identifiers

CAS 174800-02-3
Molecular Formula C27H25NO5
Molecular Weight (g/mol) 443.499
MDL Number MFCD00237023
InChI Key XGFMHBUVVWZBFT-CLOONOSVSA-N
Synonym fmoc-hyp bzl-oh, 2s,4r-1-9h-fluoren-9-yl methoxy carbonyl-4-benzyloxy pyrrolidine-2-carboxylic acid, fmoc-o-benzyl-l-4-hydroxyproline, 2s,4r-4-benzyloxy-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, ambotzfaa1790, fmoc-pro 4-obzl-oh, fmoc-hydroxyproline bzl-oh, fmoc-o-benzyl-l-4-trans-hydroxyproline, n-alpha-9-fluorenylmethoxycarbonyl-o-benzyl-l-4-hydroxyproline, 2s,4r-n-alpha-9-fluorenylmethyloxycarbonyl-4-oxybenzyl-l-proline
PubChem CID 21063752
IUPAC Name (2S,4R)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-phenylmethoxypyrrolidine-2-carboxylic acid
SMILES C1C(CN(C1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)OCC5=CC=CC=C5
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63582MD View Documents Alfa Aesar™
H63582MD
250mg Each for $40.41
 
AAH6358203 View Documents Alfa Aesar™
H6358203
1g Each for $127.36
 
AAH6358206 View Documents Alfa Aesar™
H6358206
5g Each for $527.41
 
Specifications

Chemical Identifiers

174800-02-3
443.499
XGFMHBUVVWZBFT-CLOONOSVSA-N
21063752
C1C(CN(C1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)OCC5=CC=CC=C5
C27H25NO5
MFCD00237023
fmoc-hyp bzl-oh, 2s,4r-1-9h-fluoren-9-yl methoxy carbonyl-4-benzyloxy pyrrolidine-2-carboxylic acid, fmoc-o-benzyl-l-4-hydroxyproline, 2s,4r-4-benzyloxy-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, ambotzfaa1790, fmoc-pro 4-obzl-oh, fmoc-hydroxyproline bzl-oh, fmoc-o-benzyl-l-4-trans-hydroxyproline, n-alpha-9-fluorenylmethoxycarbonyl-o-benzyl-l-4-hydroxyproline, 2s,4r-n-alpha-9-fluorenylmethyloxycarbonyl-4-oxybenzyl-l-proline
(2S,4R)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-phenylmethoxypyrrolidine-2-carboxylic acid

Specifications

174800-02-3
MFCD00237023
XGFMHBUVVWZBFT-CLOONOSVSA-N
(2S,4R)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-phenylmethoxypyrrolidine-2-carboxylic acid
21063752
Powder
White
trans-4-Benzyloxy-N-Fmoc-L-proline
C27H25NO5
fmoc-hyp bzl-oh, 2s,4r-1-9h-fluoren-9-yl methoxy carbonyl-4-benzyloxy pyrrolidine-2-carboxylic acid, fmoc-o-benzyl-l-4-hydroxyproline, 2s,4r-4-benzyloxy-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, ambotzfaa1790, fmoc-pro 4-obzl-oh, fmoc-hydroxyproline bzl-oh, fmoc-o-benzyl-l-4-trans-hydroxyproline, n-alpha-9-fluorenylmethoxycarbonyl-o-benzyl-l-4-hydroxyproline, 2s,4r-n-alpha-9-fluorenylmethyloxycarbonyl-4-oxybenzyl-l-proline
C1C(CN(C1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)OCC5=CC=CC=C5
443.499
443.49
98%
250mg
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C