missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-Benzyloxy-N-Fmoc-L-proline, 98%

Manufacturer:  Alfa Aesar™ H6358206

 View more versions of this product

Catalog No. AAH6358206


 

Specifications

Specifications

trans-4-Benzyloxy-N-Fmoc-L-proline
98%
MFCD00237023
XGFMHBUVVWZBFT-CLOONOSVSA-N
(2S,4R)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-phenylmethoxypyrrolidine-2-carboxylic acid
21063752
Powder
5g
174800-02-3
C27H25NO5
Fmoc-Hyp(Bzl)-OH
C1C(CN(C1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)OCC5=CC=CC=C5
443.499
443.49
White

Chemical Identifiers

174800-02-3
443.499
XGFMHBUVVWZBFT-CLOONOSVSA-N
21063752
C27H25NO5
MFCD00237023
Fmoc-Hyp(Bzl)-OH
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents