missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans-4-Acetamidocyclohexanol 98.0+%, TCI America™
$89.59 - $267.10
Chemical Identifiers
CAS | 27489-60-7 |
---|---|
Molecular Formula | C8H15NO2 |
Molecular Weight (g/mol) | 157.213 |
MDL Number | MFCD00051422 |
InChI Key | HWAFCRWGGRVEQL-UHFFFAOYSA-N |
PubChem CID | 90074 |
IUPAC Name | N-(4-hydroxycyclohexyl)acetamide |
SMILES | CC(=O)NC1CCC(CC1)O |
Chemical Identifiers
27489-60-7 | |
157.213 | |
HWAFCRWGGRVEQL-UHFFFAOYSA-N | |
N-(4-hydroxycyclohexyl)acetamide |
C8H15NO2 | |
MFCD00051422 | |
90074 | |
CC(=O)NC1CCC(CC1)O |
Specifications
27489-60-7 | |
C8H15NO2 | |
5 g | |
CC(=O)NC1CCC(CC1)O | |
157.213 | |
157.21 | |
Crystalline Powder |
170°C | |
MFCD00051422 | |
HWAFCRWGGRVEQL-UHFFFAOYSA-N | |
N-(4-hydroxycyclohexyl)acetamide | |
90074 | |
≥98.0% (GC) | |
trans-4-Acetamidocyclohexanol |
Safety and Handling
TSCA : No